Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Isavuconazonium sulfate | 946075-13-4 | MFCD27987905 | 97.1% | 814.84 g/mol | C35H36F2N8O9S2 | 1 ML
SDP

Catalog No. 5000214338
Encompass_Preferred

Isavuconazonium sulfate is the water-soluble prodrug of the antifungal isavuconazole, provided for research use. It is intended for biochemical and microbiological studies of invasive fungal pathogens and is supplied as a solid or as a ready-to-use 10 mM solution in DMSO, with supporting documentation for handling and quality.

  • Water-soluble prodrug of isavuconazole.
  • Suitable for in vitro studies of invasive fungal infections.
  • Available as solid and as a 10 mM solution in DMSO.
  • Purity 97.1% with COA and SDS available.
  • Storage: powder at -20 °C; solution at -80 °C for extended stability.

Catalog No. 50-002-14338 Supplier Medchemexpress LLC Supplier No. HY10037310MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Isavuconazonium sulfate is the water-soluble prodrug of the antifungal isavuconazole, provided for research use. It is intended for biochemical and microbiological studies of invasive fungal pathogens and is supplied as a solid or as a ready-to-use 10 mM solution in DMSO, with supporting documentation for handling and quality.

  • Water-soluble prodrug of isavuconazole.
  • Suitable for in vitro studies of invasive fungal infections.
  • Available as solid and as a 10 mM solution in DMSO.
  • Purity 97.1% with COA and SDS available.
  • Storage: powder at -20 °C; solution at -80 °C for extended stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.