Learn More
Abcam ISCK03, SCF/c-Kit inhibitor, 5MG
Catalog No. 502705431
Encompass_Preferred
MW 355.5 Da, Purity >99%. Cell-permeable, selective SCF/c-Kit inhibitor (IC₅₀ < 2.5 μM). Blocks SCF-induced c-kit and Erk phosphorylation without affecting HGF-induced Erk phosphorylation. Shows antiangiogenic effects. Abolishes melanin production in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5431 Supplier Abcam Supplier No. AB1450665MG
MW 355.5 Da, Purity >99%. Cell-permeable, selective SCF/c-Kit inhibitor (IC₅₀ < 2.5 μM). Blocks SCF-induced c-kit and Erk phosphorylation without affecting HGF-induced Erk phosphorylation. Shows antiangiogenic effects. Abolishes melanin production in vivo. .
The product is subject to the following: Abcam Restricted Use Statement