Learn More
ATCC Iscove's Modified Dulbecco's Medium (IMDM)
Catalog No. 502384182
Encompass
Iscove's Modified Dulbecco's Medium (IMDM) contains 4 mM L-glutamine, 4500 mg/L glucose, and 1500 mg/L sodium bicarbonate. It has applications for cell culture, cell growth, and viability.
Catalog No. 50-238-4182 Supplier ATCC Supplier No. 302005
Iscove's Modified Dulbecco's Medium (IMDM) contains 4 mM L-glutamine, 4500 mg/L glucose, and 1500 mg/L sodium bicarbonate. It has applications for cell culture, cell growth, and viability.