Learn More
FUJIFILM Biosciences Iscoves Modified Dulbeccos Med
Catalog No. 5000450252
Encompass
Iscoves Modified Dulbeccos Medium IMDM is a modification of Dulbeccos Basal Medium Eagle BME that uses high glucose and pyruvate additional amino acids 25mM HEPES selenium and other components IMDM is designed for serumfree growth of hemopoietic cells in culture and supports the growth ofLymphocytes and a variety of other hybrid cells
Catalog No. 50-004-50252 Supplier FUJIFILM Biosciences Supplier No. 9032500ML
Iscoves Modified Dulbeccos Medium IMDM is a modification of Dulbeccos Basal Medium Eagle BME that uses high glucose and pyruvate additional amino acids 25mM HEPES selenium and other components IMDM is designed for serumfree growth of hemopoietic cells in culture and supports the growth ofLymphocytes and a variety of other hybrid cells