Learn More
Abbott™ iScreen™ Dx Tox Cup, Tests 14 Drugs
Catalog No. 22911028
Change view
Click to view available options
Detectable Analytes:
Amphetamine (AMP), Barbiturates (BAR), Buprenorphine (BUP), Benzodiazepines (BZO), Cocaine (COC), Methylenedioxymethamphetamine (MDMA), Methamphetamine (mAMP), Morphine (MOP), Methadone (MTD), Oxycodone (OXY), Phencyclidine (PCP), Tricyclic Antidepressants (TCA), Cannabinoids (THC), Fentanyl (FTY), Adulteration Strips (CR, OX, PH
Catalog No. Detectable Analytes
22-911-028 Amphetamine (AMP), Barbiturates (BAR), Buprenorphine (BUP), Benzodiazepines (BZO), Cocaine (COC), Methylenedioxymethamphetamine (MDMA), Methamphetamine (mAMP), Morphine (MOP), Methadone (MTD), Oxycodone (OXY), Phencyclidine (PCP), Tricyclic Antidepressants (TCA), Cannabinoids (THC), Fentanyl (FTY), Adulteration Strips (CR, OX, PH
1 options
This item is not returnable. View return policy
Catalog No. 22-911-028 Supplier Abbott™ Supplier No. ABTDUAW114705E
Add to Cart
Add to Cart
This item is not returnable. View return policy
Detect the presence of drugs in minutes. Testing for up to 12 drugs on one device.
  • Detect the presence of drugs in minutes
  • Simple dip and read procedure
  • Room temperature storage
  • Laboratory accurate
  • 1-12 Drug configurations available
  • Results in minutes
  • 24 Month shelf life
  • CLIA Waived
Specifications
Clia Complexity Waived
Detectable Analytes Amphetamine (AMP), Barbiturates (BAR), Buprenorphine (BUP), Benzodiazepines (BZO), Cocaine (COC), Methylenedioxymethamphetamine (MDMA), Methamphetamine (mAMP), Morphine (MOP), Methadone (MTD), Oxycodone (OXY), Phencyclidine (PCP), Tricyclic Antidepressants (TCA), Cannabinoids (THC), Fentanyl (FTY), Adulteration Strips (CR, OX, PH
Sample Type Urine
Type iScreen™ Dx Tox Cup
Test Time Results in minutes