Learn More
Bio Rad Laboratories ISCRIPT CDNA SYNTHESIS KIT 25
Catalog No. NC9150725
Encompass
iScript cDNA Synthesis Kit, 25 x 20microliter reactions, Includes: 5x iScript reaction mix, iScript enzyme mixture, Nuclease-free water
Catalog No. NC9150725 Supplier Bio Rad Laboratories Supplier No. 1708890
iScript cDNA Synthesis Kit, 25 x 20microliter reactions, Includes: 5x iScript reaction mix, iScript enzyme mixture, Nuclease-free water