Learn More
Bio Rad Laboratories iScript Reverse Transcription Supermix, 5x concentration, 100 μL, 25 reactions, 20 μL reaction volume, RNase H+ MMLV reverse transcriptase, combined oligo(dT) and random primers
Catalog No. NC1646384
Encompass
iScript Reverse Transcription Supermix, 5x concentration, 100 μL, 25 reactions, 20 μL reaction volume, RNase H+ MMLV reverse transcriptase, combined oligo(dT) and random primers
Catalog No. NC1646384 Supplier Bio Rad Laboratories Supplier No. 1708840
iScript Reverse Transcription Supermix, 5x concentration, 100 μL, 25 reactions, 20 μL reaction volume, RNase H+ MMLV reverse transcriptase, combined oligo(dT) and random primers