Learn More
Bio Rad Laboratories ISCRIPT SELECT CDNA SYNTHESIS
Catalog No. 509010020
Encompass
Kit, Synthesis; Bio-Rad; iScript Select cDNA; Includes 5x iScript Select reaction mix, iScript reverse transcriptase, oligo(dT) mix, random primer mix, gene-specific primer enhancer solution, nuclease-free water; 100 x 20uL reactions
Catalog No. 50-901-0020 Supplier Bio Rad Laboratories Supplier No. 1708897
Kit, Synthesis; Bio-Rad; iScript Select cDNA; Includes 5x iScript Select reaction mix, iScript reverse transcriptase, oligo(dT) mix, random primer mix, gene-specific primer enhancer solution, nuclease-free water; 100 x 20uL reactions