Learn More
YSI 3P LifeScience ISE BUFFER SOLUTION 1000 ML
Catalog No. NC0495630
Encompass
Ammonium/Potassium ISE Buffer for use with YSI 2950 Analyzers with an ISE chamber. One 1000 mL bottle.
Catalog No. NC0495630 Supplier YSI 3P LifeScience Supplier No. 2970
Ammonium/Potassium ISE Buffer for use with YSI 2950 Analyzers with an ISE chamber. One 1000 mL bottle.