Learn More
METTLER TOLEDO 3P INDUSTRIAL ISE standard Cl | 500ml | 1000ppm
Catalog No. NC1843484
Encompass_Preferred
With this ready to use ion calibration standard no weighing and dilutiong is required anymore.
Catalog No. NC1843484 Supplier METTLER TOLEDO 3P INDUSTRIAL Supplier No. 51344772
With this ready to use ion calibration standard no weighing and dilutiong is required anymore.