Learn More
TARGETMOL CHEMICALS INC ISEPAMICIN SULFATE 50MG
SDP
Catalog No. 502579336
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Isepamicin sulfate (sch21420) Sulphate) an aminoglycoside antibacterial with good activity against strains producing type I 6-acetyltransferase inhibits bacterial protein synthesis by targeting the bacterial 30S ribosomal subunit. purity: 99%
Catalog No. 50-257-9336 Supplier TARGETMOL CHEMICALS INC Supplier No. T654950MG
Also available in 5 mg 10 mg 25 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Isepamicin sulfate (sch21420) Sulphate) an aminoglycoside antibacterial with good activity against strains producing type I 6-acetyltransferase inhibits bacterial protein synthesis by targeting the bacterial 30S ribosomal subunit. purity: 99%