Learn More
Apexbio Technology LLC Isepamicin Sulphate 67814-76-0 100mg
SDP
Catalog No. 5000093340
Encompass_Preferred
Isepamicin Sulphate (CAS 67814-76-0) is a semi-synthetic aminoglycoside antibiotic targeting the bacterial 30S ribosomal subunit It is designed to disrupt protein biosynthesis in bacteria thereby inhibiting bacterial growth Isepamicin Sulphate exerts its biological activity primarily through specific binding to the 30S ribosomal subunit interfering with bacterial protein synthesis In microbiological assays isepamicin sulfate demonstrates broad-spectrum inhibition activity with potency quantitatively evaluated by minimal inhibitory concentration (MIC) and IC50 values Based on these pharmacological properties isepamicin sulfate holds research potential in microbial resistance studies susceptibility assays and antibacterial screening involving multidrug-resistant Gram-negative pathogens
Catalog No. 50-000-93340 Supplier Apexbio Technology LLC Supplier No. B1955100
Isepamicin Sulphate (CAS 67814-76-0) is a semi-synthetic aminoglycoside antibiotic targeting the bacterial 30S ribosomal subunit It is designed to disrupt protein biosynthesis in bacteria thereby inhibiting bacterial growth Isepamicin Sulphate exerts its biological activity primarily through specific binding to the 30S ribosomal subunit interfering with bacterial protein synthesis In microbiological assays isepamicin sulfate demonstrates broad-spectrum inhibition activity with potency quantitatively evaluated by minimal inhibitory concentration (MIC) and IC50 values Based on these pharmacological properties isepamicin sulfate holds research potential in microbial resistance studies susceptibility assays and antibacterial screening involving multidrug-resistant Gram-negative pathogens