Learn More
Selleck Chemical LLC Isepamicin Sulphate S4271-25mg
SDP
Catalog No. 503110454
Encompass_Preferred
Isepamicin Sulphate (Isepamicine Isepamycin sch21420) is an aminoglycoside antibiotic which inhibits bacterial protein synthesis by targeting the bacterial 30S ribosomal subunit
Catalog No. 50-311-0454 Supplier Selleck Chemical LLC Supplier No. S427125MG
Isepamicin Sulphate (Isepamicine Isepamycin sch21420) is an aminoglycoside antibiotic which inhibits bacterial protein synthesis by targeting the bacterial 30S ribosomal subunit