Learn More
Beckman Coulter B85884, 1000 μL Extended Length Filter Tips for Biomek Automated Liquid Handlers, Sterile
Catalog No. NC2253359 Shop All Beckman Coulter Products
Encompass_Preferred
The Beckman Coulter B85884 is a pipette tip that is compatible with the Biomek series of automated liquid handlers. It has a volume range of 20-200 microliters and is made from virgin polypropylene for reliable performance. The tips have a universal fit and are guaranteed not to stick during aspiration. They are sterilized by gamma irradiation and certified as sterile and non-pyrogenic. The tips come in resealable bags of 1000 tips per bag.
This item is not returnable. View return policy
Catalog No. NC2253359 Supplier Beckman Coulter Supplier No. B85884
The Beckman Coulter B85884 is a pipette tip that is compatible with the Biomek series of automated liquid handlers. It has a volume range of 20-200 microliters and is made from virgin polypropylene for reliable performance. The tips have a universal fit and are guaranteed not to stick during aspiration. They are sterilized by gamma irradiation and certified as sterile and non-pyrogenic. The tips come in resealable bags of 1000 tips per bag.