Learn More
Selleck Chemical LLC Isethionic acid sodium salt
SDP
Catalog No. 5000003708
Encompass_Preferred
Isethionic acid sodium salt is an amphoteric detergent used in detergent bar soaps
Catalog No. 50-000-03708 Supplier Selleck Chemical LLC Supplier No. S619025MG
Isethionic acid sodium salt is an amphoteric detergent used in detergent bar soaps