Learn More
Jeio Tech Inc Jeio Tech™ ISF-7100R Floor Type Large Capacity Refrigerated Incubator Shaker (120V, 60Hz) PID Temperature Control, For High-Throughput Microbial Cultivation and Protein Expression
SDP
Catalog No. NC2310586
Encompass_Preferred
Jeio Tech ISF Refrigerating Incubator Shakers are robust, top-loading units designed to provide reliable temperature control and efficient orbital shaking for a wide range of biological and microbiological applications. The ISF series combines precise PID temperature control with a durable shaking mechanism to ensure consistent and uniform sample growth. The refrigerated model offers an extended temperature control range, enabling both cooling and heating applications. Its top-loading design allows easy access and accommodates large flasks and high-volume containers, making it suitable for high-capacity workflows. Featuring a spacious chamber, adjustable shaking speed, and forced-air circulation, the ISF Incubator Shakers offer dependable performance for research, pharmaceutical, and biotechnology laboratories requiring robust and user-friendly incubation with agitation. Shaking platform required; sold separately.
Catalog No. NC2310586 Supplier Jeio Tech Inc Supplier No. AAH23575U
Add to Cart
Add to Cart
Request a Quote
Jeio Tech ISF Refrigerating Incubator Shakers are robust, top-loading units designed to provide reliable temperature control and efficient orbital shaking for a wide range of biological and microbiological applications. The ISF series combines precise PID temperature control with a durable shaking mechanism to ensure consistent and uniform sample growth. The refrigerated model offers an extended temperature control range, enabling both cooling and heating applications. Its top-loading design allows easy access and accommodates large flasks and high-volume containers, making it suitable for high-capacity workflows. Featuring a spacious chamber, adjustable shaking speed, and forced-air circulation, the ISF Incubator Shakers offer dependable performance for research, pharmaceutical, and biotechnology laboratories requiring robust and user-friendly incubation with agitation. Shaking platform required; sold separately.