Learn More
Kuhner Shaker Inc ISF1-Z SHAKER
Catalog No. 502569935
Encompass
ISF1-Z Shaker with coolingTemperature range: ambient -15C to 80CControl of 0.1C, Accuracy +/- 0.3C at 37CUser Selectable Orbit Diameter (12.5mm, 25mm, 50mm)Loads up to 25kg (+25kg possible if required)DC direct drive with single eccentric (no belt)100% stainless steel interiorExternal Dimensions: H771 x W1275 x D845mmWeight: 210kgFIVE YEAR WARRANTY HARDWARE,1 YEAR WARRANTY ON SENSORS
Catalog No. 50-256-9935 Supplier Kuhner Shaker Inc Supplier No. SMZ1500C
Add to Cart
Add to Cart
Request a Quote
ISF1-Z Shaker with coolingTemperature range: ambient -15C to 80CControl of 0.1C, Accuracy +/- 0.3C at 37CUser Selectable Orbit Diameter (12.5mm, 25mm, 50mm)Loads up to 25kg (+25kg possible if required)DC direct drive with single eccentric (no belt)100% stainless steel interiorExternal Dimensions: H771 x W1275 x D845mmWeight: 210kgFIVE YEAR WARRANTY HARDWARE,1 YEAR WARRANTY ON SENSORS