Learn More
Kuhner Shaker Inc ISF1-ZC ECODEW
Catalog No. 502569931
Encompass
ISF1-ZC EcoDewISF1-Z with heating, shaking and humidity controlTemperature range: ambient + 10C to 80CControl of 0.1C, Accuracy +/- 0.2C at 37CAccumid humidity control to 85%r.H. Set point 1%+/-2%r.HOne-tool multi-diameter hub (3, 6, 9, 12.5, 19, 25, 50, 70mm)DC direct beltless forever-drive and high load bearingsLoads up to 60kg (application dependent)100% stainless steel interiorExternal Dimensions: H771 x W1285 x D918mmWeight: 200kgFIVE YEAR WARRANTY HARDWARE,1 YEAR WARRANTY ON SENSORS
Catalog No. 50-256-9931 Supplier Kuhner Shaker Inc Supplier No. SMZ1502
Add to Cart
Add to Cart
Request a Quote
ISF1-ZC EcoDewISF1-Z with heating, shaking and humidity controlTemperature range: ambient + 10C to 80CControl of 0.1C, Accuracy +/- 0.2C at 37CAccumid humidity control to 85%r.H. Set point 1%+/-2%r.HOne-tool multi-diameter hub (3, 6, 9, 12.5, 19, 25, 50, 70mm)DC direct beltless forever-drive and high load bearingsLoads up to 60kg (application dependent)100% stainless steel interiorExternal Dimensions: H771 x W1285 x D918mmWeight: 200kgFIVE YEAR WARRANTY HARDWARE,1 YEAR WARRANTY ON SENSORS