Learn More
Kuhner Shaker Inc ISF4-XC SHAKER INCL. HUMIDITY
Catalog No. 502570047
Encompass
ISF4-XC Shaker incl. Humidity and CO2..Temperature range: ambient - 15C to 60C..Bi-directional humidity control to 90%RH, accuracy +/-1%..Control of 0.1C, Accuracy +/- 0.3C at 37C..CO2 Control 0-20%. Control of 0.1%, Accuracy +/- 0.42% @ 20% CO2..100%...
Catalog No. 50-257-0047 Supplier Kuhner Shaker Inc Supplier No. SMX1603C
Add to Cart
Add to Cart
Request a Quote
ISF4-XC Shaker incl. Humidity and CO2..Temperature range: ambient - 15C to 60C..Bi-directional humidity control to 90%RH, accuracy +/-1%..Control of 0.1C, Accuracy +/- 0.3C at 37C..CO2 Control 0-20%. Control of 0.1%, Accuracy +/- 0.42% @ 20% CO2..100%...