Learn More
Kuhner Shaker Inc ISF4-XC SHAKER WITH COOLING..T
Catalog No. 502570049
Encompass
ISF4-XC Shaker with cooling..Temperature range: ambient - 15C to 60C..Control of 0.1C, Accuracy +/- 0.3C at 37C..CO2 Control 0-20%. Control of 0.1%, Accuracy +/- 0.42% @ 20% CO2..100% stainless steel interior..External Dimensions: H1916 x W1494 x D960m...
Catalog No. 50-257-0049 Supplier Kuhner Shaker Inc Supplier No. SMX1601C
Add to Cart
Add to Cart
Request a Quote
ISF4-XC Shaker with cooling..Temperature range: ambient - 15C to 60C..Control of 0.1C, Accuracy +/- 0.3C at 37C..CO2 Control 0-20%. Control of 0.1%, Accuracy +/- 0.42% @ 20% CO2..100% stainless steel interior..External Dimensions: H1916 x W1494 x D960m...