Learn More
Santa Cruz Biotechnology ISG15 ANTIBODY(F-9)ALEXA FLUOR
Supplier:  Santa Cruz Biotechnology SC166755 AF680
Encompass
ISG15 Antibody (F-9) Alexa Fluor® 680200 µg/mlISG15 Antibody (F-9) is a mouse monoclonal IgG1 (kappa light chain) provided at 200 µg/mlraised against amino acids 1-150 mapping at the N-terminus of ISG15 of human originrecommended for detection of ISG15 of mouse, rat and human origin by WB, IP, IF, IHC(P) and ELISA
Catalog No. NC1469807