Learn More
Enzo Life Sciences ISG15 (human), (recombinant) (His-tag) (500 µg)
SDP
Supplier:  Enzo Life Sciences BMLUW92350500
Encompass_Preferred
Mature protein for functional studies. Mature protein for functional studies. Purity: ≥95% (SDS-PAGE). Formulation: Liquid. In 20mM HEPES, pH 8.0, 50mM NaCl, 1mM DTT. Source: Produced in E. coli. Long Term Storage: -80°C.
Catalog No. 50-201-2311