Learn More
Sino Biological Recombinant Human VHL/ELOB/ELOC/CUL2/RBX1 complex Protein (Full Length, His Tag)
Supplier: Sino Biological U272380H1000
Recombinant full length human VHL, ELOB, ELOC, CUL2, and RBX1 were expressed by baculovirus in Sf9 insect cells using N-terminal His-tags for all proteins.