Learn More
Cusabio Technology LLC ISG15 ubiquitin-like modifier
Catalog No. 5000048297
Encompass_Preferred
The Mouse Ubiquitin-like protein ISG15 (ISG15) ELISA Kit is designed for the quantitative detection of ISG15 in serum plasma cell culture supernates and tissue homogenates This kit employs a sandwich ELISA method with a detection range of 0 312 ng/mL to 20 ng/mL and a sensitivity of 0 078 ng/mL ensuring precise measurement of low-abundance samples The assay requires a sample volume of 50-100 L and can be completed within 1-5 hours Results are measured at an absorbance wavelength of 450 nm providing reliable and reproducible data for research applications It is suitable for studying ISG15 expression and regulation in various biological contexts
Catalog No. 50-000-48297 Supplier Cusabio Technology LLC Supplier No. CSBEL011843MO
The Mouse Ubiquitin-like protein ISG15 (ISG15) ELISA Kit is designed for the quantitative detection of ISG15 in serum plasma cell culture supernates and tissue homogenates This kit employs a sandwich ELISA method with a detection range of 0 312 ng/mL to 20 ng/mL and a sensitivity of 0 078 ng/mL ensuring precise measurement of low-abundance samples The assay requires a sample volume of 50-100 L and can be completed within 1-5 hours Results are measured at an absorbance wavelength of 450 nm providing reliable and reproducible data for research applications It is suitable for studying ISG15 expression and regulation in various biological contexts