Learn More
Aviva Systems Biology Corp ISG20 Antibody - middle region (ARP40391_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40391_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ISG20. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ISG20. Alternate names: CD25, HEM45ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Pig, Rabbit, Rat
Catalog No. 50-014-57427