Learn More
Aviva Systems Biology Corp ISGF3G Antibody - N-terminal region (ARP31200_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31200_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ISGF3G. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human ISGF3G. Alternate names: IRF-9, ISGF3, ISGF3G, p48ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54185