Learn More
Enzo Life Sciences In situ hybridization buffer for HPV probes (Ready-to-Use) (50ml)
SDP
Catalog No. 502012504
Encompass_Preferred
In situ hybridization buffer for HPV probes (Ready-to Use) provides a high quality hybridization medium for In situ hybridization analyses using either DNA or oligonucleotide probes labeled with biotin-, or fluorescein-modified nucleotides. Long Term Storage: +4°C.
Catalog No. 50-201-2504 Supplier Enzo Life Sciences Supplier No. ENZ339050050
In situ hybridization buffer for HPV probes (Ready-to Use) provides a high quality hybridization medium for In situ hybridization analyses using either DNA or oligonucleotide probes labeled with biotin-, or fluorescein-modified nucleotides. Long Term Storage: +4°C.