Learn More
Enzo Life Sciences In situ hybridization wash reagent (30ml)
SDP
Catalog No. 502012503
Encompass_Preferred
In situ Hybridization Wash Reagent provides a high quality hybridization wash solution for in situ hybridization analyses using DNA probes labeled with biotin, digoxigenin or fluorescein. Formulation: Liquid. PBS containing deionized formamide. Long Term Storage: +4°C.
Catalog No. 50-201-2503 Supplier Enzo Life Sciences Supplier No. ENZ33809
In situ Hybridization Wash Reagent provides a high quality hybridization wash solution for in situ hybridization analyses using DNA probes labeled with biotin, digoxigenin or fluorescein. Formulation: Liquid. PBS containing deionized formamide. Long Term Storage: +4°C.