Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC ISIS 104838 | 250755-32-9 | 93.7% | 1 MG
SDP

Catalog No. 5000356006
Encompass_Preferred

ISIS 104838 is an antisense oligonucleotide agent that reduces the production of tumor necrosis factor (TNF-alpha), a substance that contributes to joint pain and swelling in rheumatoid arthritis. This product is for research use only and not sold to patients.

  • Antisense oligonucleotide agent
  • Reduces tumor necrosis factor (TNF-alpha) production
  • Aids in relieving joint pain and swelling in rheumatoid arthritis
  • Molecular weight: 7283.90
  • Appearance: Solid, white to off-white
  • Targets TNF receptor
  • Involves apoptosis pathway
  • Store at -20°C under nitrogen; in solvent, store at -80°C for 6 months or -20°C for 1 month under nitrogen.

Catalog No. 50-003-56006 Supplier Medchemexpress LLC Supplier No. HY1457261MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ISIS 104838 is an antisense oligonucleotide agent that reduces the production of tumor necrosis factor (TNF-alpha), a substance that contributes to joint pain and swelling in rheumatoid arthritis. This product is for research use only and not sold to patients.

  • Antisense oligonucleotide agent
  • Reduces tumor necrosis factor (TNF-alpha) production
  • Aids in relieving joint pain and swelling in rheumatoid arthritis
  • Molecular weight: 7283.90
  • Appearance: Solid, white to off-white
  • Targets TNF receptor
  • Involves apoptosis pathway
  • Store at -20°C under nitrogen; in solvent, store at -80°C for 6 months or -20°C for 1 month under nitrogen.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.