Learn More
Aviva Systems Biology Corp ISL1 Antibody - N-terminal region (ARP32336_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32336_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ISL1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ISL1. Alternate names: Isl-1, ISLET1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-54531