Learn More
AVANTOR MASTERFLEX (VWR LLC) Ismatec IPC-N ISM939C Digital Peristaltic Pump, Low-Speed, Adjustable Occlusion Cartridges, 24-Channel, 11.25 rpm; 115 VAC
Catalog No. NC1112394
Encompass
This multichannel programmable pump features planetary gears that drive eight stainless steel rollers in unison for smooth rotation and accurate liquid flow. The four-digit LED display shows flow rate or rpm when pump is running. Use the MAX button to instantly increase speed for priming or flushing without disturbing your programmed settings. Combine tubing sizes and formulations to suit your application. Clickngo cartridges feature automatic occlusion for simplified use, fewer tubing changes, and reproducible results. Adjustable occulsion cartridges are available separately. Install cartridges in one direction for low pulsation; in opposite direction for higher counter pressure.
Catalog No. NC1112394 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX7800041
This multichannel programmable pump features planetary gears that drive eight stainless steel rollers in unison for smooth rotation and accurate liquid flow. The four-digit LED display shows flow rate or rpm when pump is running. Use the MAX button to instantly increase speed for priming or flushing without disturbing your programmed settings. Combine tubing sizes and formulations to suit your application. Clickngo cartridges feature automatic occlusion for simplified use, fewer tubing changes, and reproducible results. Adjustable occulsion cartridges are available separately. Install cartridges in one direction for low pulsation; in opposite direction for higher counter pressure.