Learn More
Ismatec™ Replacement Cartridges
Catalog No. 13200015
Change view
Click to view available options
Material:
POM-C
Catalog No. Material
13-200-015 POM-C
1 options
Catalog No. 13-200-015 Supplier Ismatec™ Supplier No. MFLX7800195
Add to Cart
Add to Cart
Choose new cassettes based upon material of construction, tubing type and pressure capabilities
Material of construction standard POM-C or optional PVDF
Specifications
Maximum Pressure 14.5 psi
Includes Click-n-go cartridge
Material POM-C