Learn More
AVANTOR MASTERFLEX (VWR LLC) Ismatec Pump Tubing, 3-Stop, Tygon LMT-55, 1.52 mm ID; 12/PK
Catalog No. NC1537740
Encompass
The overall length of LMT-55 97619-series stop tubing is 16" (406 mm). The legacy Ismatec stop tubing was 400 mm long.
Order extension tubing under "Accessories". For technical assistance selecting tubing formulations and sizes, contact our Application Specialists.
Catalog No. NC1537740 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9761936
The overall length of LMT-55 97619-series stop tubing is 16" (406 mm). The legacy Ismatec stop tubing was 400 mm long.
Order extension tubing under "Accessories". For technical assistance selecting tubing formulations and sizes, contact our Application Specialists.