Learn More
AVANTOR MASTERFLEX (VWR LLC) Ismatec Pump Tubing, 2-Stop, Peroxide-Cured Silicone, 0.76 mm ID; 6/PK
Catalog No. NC1531346
Encompass
Appearance: Opaque Temperature: -70 to 52¿C Life at 100 rpm: 1000 hrs The first of a series of two-stop extension tubing options is below under Accessories. See all of our two-stop extension tubing formulations or contact our Application Specialists for personal assistance.
Catalog No. NC1531346 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0761624
Appearance: Opaque Temperature: -70 to 52¿C Life at 100 rpm: 1000 hrs The first of a series of two-stop extension tubing options is below under Accessories. See all of our two-stop extension tubing formulations or contact our Application Specialists for personal assistance.