Learn More
AVANTOR MASTERFLEX (VWR LLC) Pump Tubing, 3-Stop, Tygon S3™ E-Lab, 3.17 mm ID; 12/PK
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX9646149/DEL
Encompass
Ismatec SC0224-LT 3-Stop, E-Lab Tubing, 3.17mm ID, 12/pk, Designed for optimal operation in your Ismatec pumpsResistant to ozone and UV light ¿ won't crack, swell, or hardenTranslucent yellow colorTemp range: -50 to 74¿C (-58 to 165¿F)
Catalog No. NC1418939