Learn More
AVANTOR MASTERFLEX (VWR LLC) ISMTC CHEM/2001 3.2 1.6 25FT
Catalog No. 502742803
Encompass
Masterflex Ismatec Pump Tubing, Versilon™ 2001, 3.2 mm ID x 6.4 mm OD; 7.5 m, Avantor
Catalog No. 50-274-2803 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0651808
Masterflex Ismatec Pump Tubing, Versilon™ 2001, 3.2 mm ID x 6.4 mm OD; 7.5 m, Avantor