Learn More
AVANTOR MASTERFLEX (VWR LLC) ISMTC CHEM/2001 6.4 1.6 25FT
Catalog No. 502742801
Encompass
Masterflex Ismatec Pump Tubing, Versilon™ 2001, 6.4 mm ID x 9.6 mm OD; 7.5 m, Avantor
Catalog No. 50-274-2801 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0651816
Masterflex Ismatec Pump Tubing, Versilon™ 2001, 6.4 mm ID x 9.6 mm OD; 7.5 m, Avantor