Learn More
Pickering Laboratories Inc Artificial Perspiration, ISO 105-B07/105-E04, pH 8.0, Not Stabilized, each (200 mL/bottle)
SDP
Catalog No. 501354811
Encompass_Preferred
Perspiration is on everything we touch with our fingers or that comes into contact with our bodies. Sweat can react with materials and trigger allergies, or it can shorten product service life via corrosion or discoloration. Natural human perspiration has a complex composition that depends on the type of sweat gland, the person’s metabolism and hydration level. These test solutions are for all kinds of textiles and textiles in all forms. Textiles are wetted with either the acidic or alkaline solution to test the combined effect of perspiration solution and exposure to light according to ISO 105-B07, and to perspiration only according to ISO 105-E04. The pH of the acidic solution is 5.5 and that of the alkaline solution is 8.0.
Catalog No. 50-135-4811 Supplier Pickering Laboratories Inc Supplier No. 17000011
Perspiration is on everything we touch with our fingers or that comes into contact with our bodies. Sweat can react with materials and trigger allergies, or it can shorten product service life via corrosion or discoloration. Natural human perspiration has a complex composition that depends on the type of sweat gland, the person’s metabolism and hydration level. These test solutions are for all kinds of textiles and textiles in all forms. Textiles are wetted with either the acidic or alkaline solution to test the combined effect of perspiration solution and exposure to light according to ISO 105-B07, and to perspiration only according to ISO 105-E04. The pH of the acidic solution is 5.5 and that of the alkaline solution is 8.0.