Learn More
ELCOMETER INC ISO 2409 TAPE
SDP
Catalog No. NC1925627
Encompass
This tape is recommended for use with coatings up to 5 mils thick and is also acceptable for use with ISO 2409.
Catalog No. NC1925627 Supplier ELCOMETER INC Supplier No. T99993582
This tape is recommended for use with coatings up to 5 mils thick and is also acceptable for use with ISO 2409.