Learn More
TARGETMOL CHEMICALS INC ISO-BMT-124110 FORMATE 5MG
SDP
Catalog No. 502579155
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. (Iso)-BMT-124110 Formate has an inhibitory effect on the protein serine/threonine kinase AAK and is used in the treatment of Parkinson's disease schizophrenia neuropathic pain and Alzheimer's disease.
Catalog No. 50-257-9155 Supplier TARGETMOL CHEMICALS INC Supplier No. T77333L5MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. (Iso)-BMT-124110 Formate has an inhibitory effect on the protein serine/threonine kinase AAK and is used in the treatment of Parkinson's disease schizophrenia neuropathic pain and Alzheimer's disease.