Learn More
Euthanex Stand-Alone TEC 3 Isoflurane Vaporizer - Funnel Fill
Catalog No. 501540164
Encompass_Preferred
Stand-Alone TEC 3 Isoflurane Vaporizer - Funnel Fill. Construction of main vessel is brass. Used for small animals. Concentration variable between 0-5%.
Catalog No. 50-154-0164 Supplier Euthanex Supplier No. EZ155
Stand-Alone TEC 3 Isoflurane Vaporizer - Funnel Fill. Construction of main vessel is brass. Used for small animals. Concentration variable between 0-5%.