Learn More
World Precision Instrument ISO-COP-2 Carbon Monoxide Sensor - 2mm
SDP
Catalog No. 502521171
Encompass
Carbon monoxide (CO) is a versatile mediator of physiological processes. Carbon monoxide (CO) formed by internal mechanisms (endogenous) is measured in a variety of ways, but standard measurement methods are of limited utility in most biological systems.  WPI's ingenious ISO-COP-2 CO sensor measures CO in vivo or in vitro in real time!
Catalog No. 50-252-1171 Supplier World Precision Instrument Supplier No. ISOCOP2
Carbon monoxide (CO) is a versatile mediator of physiological processes. Carbon monoxide (CO) formed by internal mechanisms (endogenous) is measured in a variety of ways, but standard measurement methods are of limited utility in most biological systems.  WPI's ingenious ISO-COP-2 CO sensor measures CO in vivo or in vitro in real time!