Learn More
World Precision Instrument Iso-DAM8A Active Headstage
SDP
Supplier:  World Precision Instrument 74040
Encompass
Bridge8 is a modular, rack-mountable amplifier system based on a similar architecture to the popular ISO-DAM8A amplifier system. Bridge8 is specifically designed to be used as a signal conditioning amplifier with strain gauges and other powered transducers. Bridge8 includes differential amplifiers featuring high input impedance, high common mode rejection and low current leakage input terminals for low noise operation. The Bridge8 also features a half bridge switch and channel offset A wide variety of WPI transducers are available for force, temperature, pressure and light measurements. The Bridge8 amplifier is a clear choice where convenience and quality are required. Any combination of ISO-DAM8 or Bridge8 modules may be combined in the same enclosure. The ISO-DAM8A module is also available in this rack configuration.
Catalog No. 50-252-9297