Learn More
TARGETMOL CHEMICALS INC ISO-FOSDEVIRINE 5MG
SDP
Catalog No. 502580346
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg and bulk. Please contact Fisher for quotes. (Iso)-Fosdevirine ( (Iso)-GSK2248761) is a selective and potent non-nucleoside reverse transcriptase (NNRTI) inhibitor with anti-HIV activity for the study of neurological disorders such as late-onset epilepsy. purity: 99%
Catalog No. 50-258-0346 Supplier TARGETMOL CHEMICALS INC Supplier No. T31860L5MG
Add to Cart
Add to Cart
Also available in 1 mg 10 mg 25 mg 50 mg and bulk. Please contact Fisher for quotes. (Iso)-Fosdevirine ( (Iso)-GSK2248761) is a selective and potent non-nucleoside reverse transcriptase (NNRTI) inhibitor with anti-HIV activity for the study of neurological disorders such as late-onset epilepsy. purity: 99%