Learn More
Medchemexpress LLC Iso-H7 dihydrochloride | 140663-38-3 | 99.1% | 364.29 | 1 MG
SDP
Catalog No. 5000435797
Encompass_Preferred
Iso-H7 dihydrochloride is a protein kinase inhibitor. It exhibits IC50 values of 22 μM for Protein Kinase C (PKC) and 34 μM for Protein Kinase A (PKA). This compound has been observed to have a negative control effect on PKA in the context of satellite cell myogenesis.
  • Potent inhibitor of protein kinases, specifically PKC and PKA.
  • Demonstrates inhibitory activity against PKC with an IC50 of 22 μM.
  • Inhibits PKA with an IC50 of 34 μM.
  • Exerts negative control over PKA's role in satellite cell myogenesis.
  • Suitable for research applications.
Catalog No. 50-004-35797 Supplier Medchemexpress LLC Supplier No. HY1263761MG
Iso-H7 dihydrochloride is a protein kinase inhibitor. It exhibits IC50 values of 22 μM for Protein Kinase C (PKC) and 34 μM for Protein Kinase A (PKA). This compound has been observed to have a negative control effect on PKA in the context of satellite cell myogenesis.
  • Potent inhibitor of protein kinases, specifically PKC and PKA.
  • Demonstrates inhibitory activity against PKC with an IC50 of 22 μM.
  • Inhibits PKA with an IC50 of 34 μM.
  • Exerts negative control over PKA's role in satellite cell myogenesis.
  • Suitable for research applications.