Learn More
Medchemexpress LLC Iso-H7 dihydrochloride | 140663-38-3 | 99.1% | 364.29 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY12637610MM/1ML
Encompass_Preferred
Iso-H7 dihydrochloride is a potent protein kinase inhibitor. It is characterized by its inhibitory activity against PKC and PKA, demonstrating IC50 values of 22 μM and 34 μM, respectively. This compound also plays a role in the negative control of PKA on satellite cell myogenesis.
  • Inhibits protein kinase activity
  • Effective against PKC with an IC50 of 22 μM
  • Effective against PKA with an IC50 of 34 μM
  • Exhibits negative control of PKA on satellite cell myogenesis
Catalog No. 50-004-38063