Learn More
Medchemexpress LLC Iso-H7 (dihydrochloride) | 140663-38-3 | 99.1% | 364.29 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY12637610MG
Encompass_Preferred
Iso-H7 dihydrochloride is a protein kinase inhibitor that exhibits IC50 values of 22 and 34 μM for PKC and PKA, respectively. It also acts as a negative control for PKA in satellite cell myogenesis. This product is intended for research use only and is not sold to patients.
  • Inhibitor of protein kinase.
  • Specifically inhibits PKC with an IC50 of 22 μM and PKA with an IC50 of 34 μM.
  • Demonstrates negative control of PKA on satellite cell myogenesis.
Catalog No. 50-004-38195