Learn More
Medchemexpress LLC Iso-H7 dihydrochloride | 140663-38-3 | 99.1% | 364.29 | 25 MG
SDP
Catalog No. 5000440065
Encompass_Preferred
Iso-H7 dihydrochloride is an inhibitor of protein kinase with IC50 values of 22 and 34 μM for PKC and PKA, respectively. It has a negative control effect on PKA in satellite cell myogenesis.
  • Protein kinase inhibitor.
  • Inhibits PKC (IC50 = 22 μM).
  • Inhibits PKA (IC50 = 34 μM).
  • Negative control of PKA on satellite cell myogenesis.
Catalog No. 50-004-40065 Supplier Medchemexpress LLC Supplier No. 25MGHY126376
Iso-H7 dihydrochloride is an inhibitor of protein kinase with IC50 values of 22 and 34 μM for PKC and PKA, respectively. It has a negative control effect on PKA in satellite cell myogenesis.
  • Protein kinase inhibitor.
  • Inhibits PKC (IC50 = 22 μM).
  • Inhibits PKA (IC50 = 34 μM).
  • Negative control of PKA on satellite cell myogenesis.