Learn More
Medchemexpress LLC Iso-H7 dihydrochloride | 140663-38-3 | MFCD00132899 | 99.1% | 364.29 g/mol | C14H19Cl2N3O2S | 5 MG
SDP
Catalog No. 5000264529
Encompass_Preferred
Iso-H7 dihydrochloride is a small-molecule protein kinase inhibitor supplied as the dihydrochloride salt for research use. It is intended for biochemical and cell-based studies to modulate kinase activity and is provided in small, high-purity quantities for assay screening and optimization.
  • High purity for reliable experimental results.
  • Dihydrochloride salt form to aid solubility in polar solvents.
  • Available in small milligram pack sizes suitable for screening.
  • Validated for use as a protein kinase inhibitor in biochemical and cellular assays.
  • Accompanied by datasheet, certificate of analysis, and safety data sheet for quality and safety information.
Catalog No. 50-002-64529 Supplier Medchemexpress LLC Supplier No. HY1263765MG
Iso-H7 dihydrochloride is a small-molecule protein kinase inhibitor supplied as the dihydrochloride salt for research use. It is intended for biochemical and cell-based studies to modulate kinase activity and is provided in small, high-purity quantities for assay screening and optimization.
  • High purity for reliable experimental results.
  • Dihydrochloride salt form to aid solubility in polar solvents.
  • Available in small milligram pack sizes suitable for screening.
  • Validated for use as a protein kinase inhibitor in biochemical and cellular assays.
  • Accompanied by datasheet, certificate of analysis, and safety data sheet for quality and safety information.