Learn More
Mercodia Inc Iso-Insulin ELISA, Human, Total Insulin Measurement, Serum Plasma, 96 Wells
SDP
Supplier:  Mercodia Inc 10112801
Encompass_Preferred
A high quality enzyme immunoassay for the quantification of insulin in serum or plasma. Designed to measure total insulin: combination of endogenous insulin and insulin analogs. Can be combined with the highly specific Mercodia Human Insulin ELISA to enable the calculation of insulin analog concentrations by subtracting endogenous insulin. Cross reacts with insulin from a broad range of species.
Catalog No. NC9936189